Anti-TNFSF13B Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TNFSF13B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77808-912
- 77808-913
- Antigen Symbol:
- TNFSF13B
- TNFSF13B
- Antigen Name:
- Tumor necrosis factor ligand superfamily member 13B protein (68-285AA)
- Tumor necrosis factor ligand superfamily member 13B protein (68-285AA)
- Antigen Synonyms:
- ApoL related ligand TALL 1|B cell Activating Factor|B lymphocyte stimulator|B-cell-activating factor|BAFF|BLyS|CD 257|CD257|CD257 antigen|Delta BAFF|Dendritic cell derived TNF like molecule|Dendritic cell-derived TNF-like molecule|DTL|DTL precursor|PRO738|soluble form|TALL 1|TALL-1|TALL1|THANK|TN13B_HUMAN|TNF and APOL related leukocyte expressed ligand 1|TNF homolog that activates apoptosis|TNF homolog that activates apoptosis NKFB and JNK.|TNF- and APOL-related leukocyte expressed ligand 1|TNFSF13B|TNFSF20|TNLG7A|Tumor necrosis factor (ligand) superfamily member 13b|Tumor necrosis factor ligand 7A|Tumor necrosis factor ligand superfamily member 13b|Tumor necrosis factor ligand superfamily member 20|Tumor necrosis factor like protein ZTNF4|Tumor necrosis factor superfamily member 13B|UNQ401|ZTNF4
- ApoL related ligand TALL 1|B cell Activating Factor|B lymphocyte stimulator|B-cell-activating factor|BAFF|BLyS|CD 257|CD257|CD257 antigen|Delta BAFF|Dendritic cell derived TNF like molecule|Dendritic cell-derived TNF-like molecule|DTL|DTL precursor|PRO738|soluble form|TALL 1|TALL-1|TALL1|THANK|TN13B_HUMAN|TNF and APOL related leukocyte expressed ligand 1|TNF homolog that activates apoptosis|TNF homolog that activates apoptosis NKFB and JNK.|TNF- and APOL-related leukocyte expressed ligand 1|TNFSF13B|TNFSF20|TNLG7A|Tumor necrosis factor (ligand) superfamily member 13b|Tumor necrosis factor ligand 7A|Tumor necrosis factor ligand superfamily member 13b|Tumor necrosis factor ligand superfamily member 20|Tumor necrosis factor like protein ZTNF4|Tumor necrosis factor superfamily member 13B|UNQ401|ZTNF4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y275
- Q9Y275
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together