Anti-CROT Rabbit Polyclonal Antibody
Catalog # 77806-263
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the Internal region of CROT.
- Antigen Symbol:CROT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UKG9
- Antigen Synonyms:Carnitine O octanoyltransferase|COT|CROT|OCTC_HUMAN|Peroxisomal carnitine acyltransferase|Peroxisomal carnitine O octanoyltransferase|Peroxisomal carnitine O-octanoyltransferase
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: CROT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat