Anti-RNF113B Rabbit Polyclonal Antibody
77820-738
New Product
- Antigen Name:Synthesized peptide derived from Internal of RNF113B.
- Antigen Symbol:RNF113B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8IZP6
- Antigen Synonyms:RNF113B|RNF161|ZNF183L1|RING finger protein 113B|Zinc finger protein 183-like 1
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: RNF113B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human