Anti-TXNDC9 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TXNDC9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-753
- 77803-753
- Antigen Symbol:
- TXNDC9
- TXNDC9
- Antigen Name:
- Thioredoxin domain-containing protein 9 protein (1-188AA)
- Thioredoxin domain-containing protein 9 protein (1-188AA)
- Antigen Synonyms:
- APACD|ATP binding protein associated with cell differentiation|ATP-binding protein associated with cell differentiation|ES cell related protein|PHLP3|Phosducin like protein 3|Protein 1 4|Protein 1-4|Thioredoxin domain containing 9|Thioredoxin domain containing protein 9|Thioredoxin domain-containing protein 9|TXND9_HUMAN|TXNDC9 protein
- APACD|ATP binding protein associated with cell differentiation|ATP-binding protein associated with cell differentiation|ES cell related protein|PHLP3|Phosducin like protein 3|Protein 1 4|Protein 1-4|Thioredoxin domain containing 9|Thioredoxin domain containing protein 9|Thioredoxin domain-containing protein 9|TXND9_HUMAN|TXNDC9 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14530
- O14530
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together