Anti-EPPIN Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: EPPIN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77800-864
- 77800-863
- Antigen Symbol:
- EPPIN
- EPPIN
- Antigen Name:
- Eppin protein (22-133AA)
- Eppin protein (22-133AA)
- Antigen Synonyms:
- Cancer/testis antigen 71|CT 71|CT71|dJ461P17.2|Epididymal protease inhibitor|EPPI_HUMAN|EPPIN 1|EPPIN 2|EPPIN 3|EPPIN1|EPPIN2|EPPIN3|Protease inhibitor WAP7|Serine peptidase inhibitor like with Kunitz and WAP domains 1 (eppin)|Serine peptidase inhibitor like with Kunitz and WAP domains 1|Serine protease inhibitor-like with Kunitz and WAP domains 1|SPINLW 1|WAP 7|WAP four disulfide core domain 7|WAP four disulfide core domain protein 7|WAP four-disulfide core domain protein 7|WAP7|WFDC 7|WFDC7
- Cancer/testis antigen 71|CT 71|CT71|dJ461P17.2|Epididymal protease inhibitor|EPPI_HUMAN|EPPIN 1|EPPIN 2|EPPIN 3|EPPIN1|EPPIN2|EPPIN3|Protease inhibitor WAP7|Serine peptidase inhibitor like with Kunitz and WAP domains 1 (eppin)|Serine peptidase inhibitor like with Kunitz and WAP domains 1|Serine protease inhibitor-like with Kunitz and WAP domains 1|SPINLW 1|WAP 7|WAP four disulfide core domain 7|WAP four disulfide core domain protein 7|WAP four-disulfide core domain protein 7|WAP7|WFDC 7|WFDC7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95925
- O95925
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together