Anti-EIF3A Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EIF3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-296
- 77825-297
- Antigen Symbol:
- EIF3A
- EIF3A
- Antigen Name:
- Eukaryotic translation initiation factor 3 subunit A protein (1288-1379AA)
- Eukaryotic translation initiation factor 3 subunit A protein (1288-1379AA)
- Antigen Synonyms:
- centrosomin homolog|cytoplasmic protein p167|eIF 3 theta|eIF-3-theta|EIF3|eIF3 p167|eIF3 p170|eIF3 p180|EIF3 p180 subunit|eIF3 p185|eIF3 theta|eIF3a|EIF3A_HUMAN|EIF3S10|eukaryotic translation initiation factor 3 subunit 10 (theta 150 170kD)|eukaryotic translation initiation factor 3 subunit 10 (theta 170kD)|eukaryotic translation initiation factor 3 subunit 10 170kDa|Eukaryotic translation initiation factor 3 subunit 10|eukaryotic translation initiation factor 3 subunit 10 theta 150 170kDa|Eukaryotic translation initiation factor 3 subunit A|OTTHUMP00000020589|P167|p180|p185|TIF32
- centrosomin homolog|cytoplasmic protein p167|eIF 3 theta|eIF-3-theta|EIF3|eIF3 p167|eIF3 p170|eIF3 p180|EIF3 p180 subunit|eIF3 p185|eIF3 theta|eIF3a|EIF3A_HUMAN|EIF3S10|eukaryotic translation initiation factor 3 subunit 10 (theta 150 170kD)|eukaryotic translation initiation factor 3 subunit 10 (theta 170kD)|eukaryotic translation initiation factor 3 subunit 10 170kDa|Eukaryotic translation initiation factor 3 subunit 10|eukaryotic translation initiation factor 3 subunit 10 theta 150 170kDa|Eukaryotic translation initiation factor 3 subunit A|OTTHUMP00000020589|P167|p180|p185|TIF32
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14152
- Q14152
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together