Anti-ZDHHC2 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZDHHC2
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-558
- 77841-557
- Antigen Symbol:
- ZDHHC2
- ZDHHC2
- Antigen Name:
- Palmitoyltransferase ZDHHC2 protein (230-367AA)
- Palmitoyltransferase ZDHHC2 protein (230-367AA)
- Antigen Synonyms:
- ZDHHC2|REAM|REC|ZNF372|Palmitoyltransferase ZDHHC2|EC 2.3.1.225|Reduced expression associated with metastasis protein|Reduced expression in cancer protein|Zinc finger DHHC domain-containing protein 2|DHHC-2|Zinc finger protein 372
- ZDHHC2|REAM|REC|ZNF372|Palmitoyltransferase ZDHHC2|EC 2.3.1.225|Reduced expression associated with metastasis protein|Reduced expression in cancer protein|Zinc finger DHHC domain-containing protein 2|DHHC-2|Zinc finger protein 372
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UIJ5
- Q9UIJ5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together