To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Gem-associated protein 2 protein (1-280AA)
- Antigen Symbol:GEMIN2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O14893
- Antigen Synonyms:Component of gems 2|Gem (nuclear organelle) associated protein 2|Gem associated protein 2|GEMI2_HUMAN|Gemin-2|gemin2|SIP 1|SIP-1|SIP1|SIP1 delta|SIP1-delta|SMN interacting protein 1|SMN interacting protein 1 delta|SMN-interacting protein 1|Survival interacting protein 1|Survival of motor neuron protein interacting protein 1|Survival of motor neuron protein-interacting protein 1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended Dilution: WB: 1:500 to 1:5000, IF: 1:50 to 1:200, IP: 1:200 to 1:2000
Type:
Antigen: GEMIN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human