Anti-MAP3K21 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAP3K21
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-232
- 77828-233
- Antigen Symbol:
- MAP3K21
- MAP3K21
- Antigen Name:
- Mitogen-activated protein kinase kinase kinase 21 protein (640-794AA)
- Mitogen-activated protein kinase kinase kinase 21 protein (640-794AA)
- Antigen Synonyms:
- dJ862P8.3|KIAA1804|M3KL4_HUMAN|Mitogen activated protein kinase kinase kinase|Mitogen-activated protein kinase kinase kinase MLK4|Mixed lineage kinase 4|MLK 4|Mlk4|RP5 862P8.2
- dJ862P8.3|KIAA1804|M3KL4_HUMAN|Mitogen activated protein kinase kinase kinase|Mitogen-activated protein kinase kinase kinase MLK4|Mixed lineage kinase 4|MLK 4|Mlk4|RP5 862P8.2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5TCX8
- Q5TCX8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together