Anti-ATP2C1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: ATP2C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77820-902
- 77820-901
- Antigen Symbol:
- ATP2C1
- ATP2C1
- Antigen Name:
- Fusion protein of ATP2C1
- Fusion protein of ATP2C1
- Antigen Synonyms:
- 1700121J11Rik|AT2C1_HUMAN|ATP dependent Ca(2+) pump PMR1|ATP-dependent Ca(2+) pump PMR1|ATP2C1|ATP2C1A|ATPase 2C1|ATPase Ca(2+) sequestering|ATPase Ca++ transporting type 2C member 1|AW061228|BCPM|Calcium transporting ATPase type 2C member 1|Calcium-transporting ATPase type 2C member 1|D930003G21Rik|HHD|hSPCA1|HUSSY 28|KIAA1347|MGC58010|MGC93231|OTTHUMP00000216066|OTTHUMP00000216068|OTTHUMP00000216069|OTTHUMP00000216071|OTTHUMP00000216072|OTTHUMP00000216073|OTTHUMP00000216074|OTTHUMP00000216075|PMR1|rat|homolog of|PMR1L|Secretory pathway Ca(2+) ATPase 1|Secretory pathway Ca(2+)-transporting ATPase|Secretory pathway Ca2+/Mn2+ ATPase 1|SPCA|SPCA1
- 1700121J11Rik|AT2C1_HUMAN|ATP dependent Ca(2+) pump PMR1|ATP-dependent Ca(2+) pump PMR1|ATP2C1|ATP2C1A|ATPase 2C1|ATPase Ca(2+) sequestering|ATPase Ca++ transporting type 2C member 1|AW061228|BCPM|Calcium transporting ATPase type 2C member 1|Calcium-transporting ATPase type 2C member 1|D930003G21Rik|HHD|hSPCA1|HUSSY 28|KIAA1347|MGC58010|MGC93231|OTTHUMP00000216066|OTTHUMP00000216068|OTTHUMP00000216069|OTTHUMP00000216071|OTTHUMP00000216072|OTTHUMP00000216073|OTTHUMP00000216074|OTTHUMP00000216075|PMR1|rat|homolog of|PMR1L|Secretory pathway Ca(2+) ATPase 1|Secretory pathway Ca(2+)-transporting ATPase|Secretory pathway Ca2+/Mn2+ ATPase 1|SPCA|SPCA1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P98194
- P98194
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together