Anti-LRRN3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LRRN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-625
- 77840-624
- Antigen Symbol:
- LRRN3
- LRRN3
- Antigen Name:
- Leucine-rich repeat neuronal protein 3 protein (201-500AA)
- Leucine-rich repeat neuronal protein 3 protein (201-500AA)
- Antigen Synonyms:
- LRRN3|Nbla10363|UNQ194/PRO220|Leucine-rich repeat neuronal protein 3|Neuronal leucine-rich repeat protein 3|NLRR-3
- LRRN3|Nbla10363|UNQ194/PRO220|Leucine-rich repeat neuronal protein 3|Neuronal leucine-rich repeat protein 3|NLRR-3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H3W5
- Q9H3W5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together