Anti-TOMM40 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TOMM40
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-313
- 77803-313
- Antigen Symbol:
- TOMM40
- TOMM40
- Antigen Name:
- Mitochondrial import receptor subunit TOM40 homolog protein (1-361AA)
- Mitochondrial import receptor subunit TOM40 homolog protein (1-361AA)
- Antigen Synonyms:
- C19orf1|D19S1177E|Haymaker protein|Mitochondrial import receptor subunit TOM40 homolog|Mitochondrial outer membrane protein|Mitochondrial outer membrane protein TOM40|p38.5|PER EC1|PEREC1|Probable mitochondrial import receptor subunit TOM40 homolog|Protein Haymaker|TOM40|TOM40_HUMAN|TOMM40|Translocase of outer membrane 40 kDa subunit homolog|Translocase of outer mitochondrial membrane 40|Translocase of outer mitochondrial membrane 40 homolog (yeast)|Translocase of outer mitochondrial membrane 40 homolog|Translocase of outer mitochondrial membrane 40, yeast, homolog of
- C19orf1|D19S1177E|Haymaker protein|Mitochondrial import receptor subunit TOM40 homolog|Mitochondrial outer membrane protein|Mitochondrial outer membrane protein TOM40|p38.5|PER EC1|PEREC1|Probable mitochondrial import receptor subunit TOM40 homolog|Protein Haymaker|TOM40|TOM40_HUMAN|TOMM40|Translocase of outer membrane 40 kDa subunit homolog|Translocase of outer mitochondrial membrane 40|Translocase of outer mitochondrial membrane 40 homolog (yeast)|Translocase of outer mitochondrial membrane 40 homolog|Translocase of outer mitochondrial membrane 40, yeast, homolog of
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O96008
- O96008
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together