Anti-ARHGAP1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ARHGAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
- Catalog No:
- 77827-109
- 77827-110
- Antigen Symbol:
- ARHGAP1
- ARHGAP1
- Antigen Name:
- Rho GTPase-activating protein 1 protein (216-308AA)
- Rho GTPase-activating protein 1 protein (216-308AA)
- Antigen Synonyms:
- ARHGAP 1|Arhgap1|CDC42 GTPase activating protein|Cdc42 GTPase-activating protein|CDC42GAP|GTPase activating protein rhoOGAP|GTPase-activating protein rhoOGAP|GTPase-activating protein, Rho, 1|OTTHUMP00000233485|OTTHUMP00000233486|p50 RhoGAP|p50-RhoGAP|p50rhoGAP|RHG01_HUMAN|Rho GTPase activating protein 1|Rho GTPase-activating protein 1|Rho related small GTPase protein activator|Rho type GTPase activating protein 1|Rho-related small GTPase protein activator|Rho-type GTPase-activating protein 1|RHOGAP|RHOGAP1
- ARHGAP 1|Arhgap1|CDC42 GTPase activating protein|Cdc42 GTPase-activating protein|CDC42GAP|GTPase activating protein rhoOGAP|GTPase-activating protein rhoOGAP|GTPase-activating protein, Rho, 1|OTTHUMP00000233485|OTTHUMP00000233486|p50 RhoGAP|p50-RhoGAP|p50rhoGAP|RHG01_HUMAN|Rho GTPase activating protein 1|Rho GTPase-activating protein 1|Rho related small GTPase protein activator|Rho type GTPase activating protein 1|Rho-related small GTPase protein activator|Rho-type GTPase-activating protein 1|RHOGAP|RHOGAP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q07960
- Q07960
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together