Anti-RAI14 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAI14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-659
- 77837-658
- Antigen Symbol:
- RAI14
- RAI14
- Antigen Name:
- Ankycorbin protein (242-346AA)
- Ankycorbin protein (242-346AA)
- Antigen Synonyms:
- Ankycorbin|Ankyrin repeat and coiled coil structure containing protein|Ankyrin repeat and coiled-coil structure-containing protein|DKFZp564G013|KIAA1334|NORPEG|Novel retinal pigment epithelial cell protein|RAI13|RAI14|RAI14_HUMAN|Retinoic acid induced 14|Retinoic acid induced protein 14|Retinoic acid-induced protein 14
- Ankycorbin|Ankyrin repeat and coiled coil structure containing protein|Ankyrin repeat and coiled-coil structure-containing protein|DKFZp564G013|KIAA1334|NORPEG|Novel retinal pigment epithelial cell protein|RAI13|RAI14|RAI14_HUMAN|Retinoic acid induced 14|Retinoic acid induced protein 14|Retinoic acid-induced protein 14
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P0K7
- Q9P0K7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together