Anti-PICALM Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PICALM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-751
- 77824-750
- Antigen Symbol:
- PICALM
- PICALM
- Antigen Name:
- Phosphatidylinositol-binding clathrin assembly protein (517-624AA)
- Phosphatidylinositol-binding clathrin assembly protein (517-624AA)
- Antigen Synonyms:
- CALM|Clathrin assembly lymphoid myeloid leukemia|Clathrin assembly lymphoid myeloid leukemia protein|CLTH|LAP|Phosphatidylinositol-binding clathrin assembly protein|PICAL_HUMAN|PICALM
- CALM|Clathrin assembly lymphoid myeloid leukemia|Clathrin assembly lymphoid myeloid leukemia protein|CLTH|LAP|Phosphatidylinositol-binding clathrin assembly protein|PICAL_HUMAN|PICALM
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13492
- Q13492
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together