Anti-THOC1 Rabbit Polyclonal Antibody
Catalog # 77802-504
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:THOC1_HUMAN
- Antigen Symbol:THOC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96FV9
- Antigen Synonyms:hTREX84|Death domain containing protein p84N5|HPR 1|HPR1|Nuclear matrix protein p84|P84|p84N5|Tho 1|THO complex 1|THO complex subunit 1|Tho1|THOC 1|Thoc1|THOC1_HUMAN
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: THOC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat