Anti-TFB2M Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TFB2M
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-195
- 77837-194
- Antigen Symbol:
- TFB2M
- TFB2M
- Antigen Name:
- Dimethyladenosine transferase 2, mitochondrial protein (30-108AA)
- Dimethyladenosine transferase 2, mitochondrial protein (30-108AA)
- Antigen Synonyms:
- N''-adenosyl(rRNA) dimethyltransferase 2|S adenosylmethionine 6 N', N' adenosyl(rRNA) dimethyltransferase 2|Dimethyladenosine transferase 2|dimethyladenosine transferase 2 mitochondrial|FLJ22661|FLJ23182|h mtTFB|h mtTFB2|h-mtTFB|h-mtTFB2|HCV NS5A transactivated protein 5|HCV NS5A-transactivated protein 5|hepatitis C virus NS5A transactivated protein 5|Hepatitis C virus NS5A-transactivated protein 5|Hkp1|hTFB2M|Mitochondrial 12S rRNA dimethylase 2|mitochondrial|Mitochondrial dimethyladenosine transferase 2|Mitochondrial transcription factor B2|mtTFB|mtTFB2|NS5ATP5|S-adenosylmethionine-6-N''|TFB2M|TFB2M_HUMAN|Transcription Factor B2|Transcription factor B2 mitochondrial
- N''-adenosyl(rRNA) dimethyltransferase 2|S adenosylmethionine 6 N', N' adenosyl(rRNA) dimethyltransferase 2|Dimethyladenosine transferase 2|dimethyladenosine transferase 2 mitochondrial|FLJ22661|FLJ23182|h mtTFB|h mtTFB2|h-mtTFB|h-mtTFB2|HCV NS5A transactivated protein 5|HCV NS5A-transactivated protein 5|hepatitis C virus NS5A transactivated protein 5|Hepatitis C virus NS5A-transactivated protein 5|Hkp1|hTFB2M|Mitochondrial 12S rRNA dimethylase 2|mitochondrial|Mitochondrial dimethyladenosine transferase 2|Mitochondrial transcription factor B2|mtTFB|mtTFB2|NS5ATP5|S-adenosylmethionine-6-N''|TFB2M|TFB2M_HUMAN|Transcription Factor B2|Transcription factor B2 mitochondrial
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H5Q4
- Q9H5Q4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together