Anti-TBX18 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:10 to 1:50
Type:
Antigen: TBX18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77821-213
- 77821-212
- Antigen Symbol:
- TBX18
- TBX18
- Antigen Name:
- Fusion protein of TBX18
- Fusion protein of TBX18
- Antigen Synonyms:
- T box 18|T box protein 18|T box transcription factor TBX18|T-box protein 18|T-box transcription factor TBX18|TBX18|TBX18_HUMAN
- T box 18|T box protein 18|T box transcription factor TBX18|T-box protein 18|T-box transcription factor TBX18|TBX18|TBX18_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95935
- O95935
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together