Anti-SPIN2B Rabbit Polyclonal Antibody
Catalog # 77800-853
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:SPIN-2B
- Antigen Symbol:SPIN2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9BPZ2
- Antigen Synonyms:SPIN2B|SPIN2|Spindlin-2B|Spindlin-like protein 2B|SPIN-2|SPIN-2B
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SPIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat