Anti-PFKL Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: PFKL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77793-865
- 77793-866
- Antigen Symbol:
- PFKL
- PFKL
- Antigen Name:
- ATP-dependent 6-phosphofructokinase, liver type protein (229-479AA)
- ATP-dependent 6-phosphofructokinase, liver type protein (229-479AA)
- Antigen Synonyms:
- PFKL| ATP-dependent 6-phosphofructokinase| liver type| ATP-PFK| PFK-L| EC 2.7.1.11| 6-phosphofructokinase type B| Phosphofructo-1-kinase isozyme B| PFK-B| Phosphohexokinase
- PFKL| ATP-dependent 6-phosphofructokinase| liver type| ATP-PFK| PFK-L| EC 2.7.1.11| 6-phosphofructokinase type B| Phosphofructo-1-kinase isozyme B| PFK-B| Phosphohexokinase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17858
- P17858
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together