Anti-RABEPK Rabbit Polyclonal Antibody
Catalog # 77796-510
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:RP23-446N16.2
- Antigen Symbol:RABEPK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q7Z6M1
- Antigen Synonyms:40 kDa Rab9 effector protein|8430412M01Rik|9530020d24rik|AV073337|bA65N13.1|C87311|DKFZp686P1077|OTTHUMP00000022126|OTTMUSP00000012898|OTTMUSP00000012899|OTTMUSP00000041318|p40|RAB9 effector p40|Rab9 effector protein with Kelch motifs|RAB9p40|RABEK_HUMAN|RABEPK|RABEPK protein|RGD1310612|RP11 65N13.1|RP23-446N16.2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: RABEPK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat