Anti-RAB38 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAB38
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-401
- 77796-402
- Antigen Symbol:
- RAB38
- RAB38
- Antigen Name:
- Ras-related protein Rab-38 protein (1-211AA)
- Ras-related protein Rab-38 protein (1-211AA)
- Antigen Synonyms:
- Antigen NY MEL 1|Melanoma antigen NY MEL 1|Melanoma antigen NY-MEL-1|NY MEL 1|RAB 38|Rab related GTP binding protein|Rab38|RAB38 member RAS oncogene family|RAB38_HUMAN|Ras related protein Rab 38|Ras-related protein Rab-38|rrGTPbp
- Antigen NY MEL 1|Melanoma antigen NY MEL 1|Melanoma antigen NY-MEL-1|NY MEL 1|RAB 38|Rab related GTP binding protein|Rab38|RAB38 member RAS oncogene family|RAB38_HUMAN|Ras related protein Rab 38|Ras-related protein Rab-38|rrGTPbp
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P57729
- P57729
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together