Anti-mgsA Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: mgsA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacillus cereus
Specifications
- Catalog No:
- 77818-998
- 77818-999
- Antigen Symbol:
- mgsA
- mgsA
- Antigen Name:
- Recombinant Bacillus cereus Methylglyoxal synthase protein (1-131AA)
- Recombinant Bacillus cereus Methylglyoxal synthase protein (1-131AA)
- Antigen Synonyms:
- BCA_1592Methylglyoxal synthase|MGS|EC 4.2.3.3
- BCA_1592Methylglyoxal synthase|MGS|EC 4.2.3.3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Bacillus cereus
- Bacillus cereus
- Host:
- Rabbit
- Rabbit
- Gene ID:
- C1EN30
- C1EN30
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together