Anti-ITSN1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77824-547
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Intersectin-1 protein (773-916AA)
- Antigen Symbol:ITSN1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q15811
- Antigen Synonyms:Human intersectin SH3 domain containing protein SH3P17|Intersectin 1 SH3 domain protein|intersectin 1 short form variant 3|intersectin 1 short form variant, 11|Intersectin short form|intersectin short variant 12|Intersectin-1|Intersectin1|ITSN 1|ITSN|ITSN1_HUMAN|MGC134948|MGC134949|OTTHUMP00000067855|OTTHUMP00000067856|OTTHUMP00000067857|OTTHUMP00000194810|OTTHUMP00000194811|OTTHUMP00000194812|OTTHUMP00000206688|SH3 domain containing protein 1A|SH3 domain protein 1A|SH3 domain-containing protein 1A|SH3D1A|SH3P17|Src homology 3 domain containing protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500
Type:
Antigen: ITSN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human