Anti-SPDL1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: SPDL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-625
- 77842-626
- Antigen Symbol:
- SPDL1
- SPDL1
- Antigen Name:
- Synthetic peptide of SPDL1
- Synthetic peptide of SPDL1
- Antigen Synonyms:
- AA409762| Arsenite related gene 1 protein| Arsenite-related gene 1 protein| CCDC 99| Ccdc99| Coiled coil domain containing 99| Coiled-coil domain-containing protein 99| FLJ20364| FLJ40690| hSpindly| MGC124977| Protein Spindly| rhabdomyosarcoma antigen MU RMS 40.4A| Rhabdomyosarcoma antigen MU-RMS-40.4A| Rrhabdomyosarcoma antigen protein MU RMS 40.4A| SPDL1| SPDLY_HUMAN| Spindle apparatus coiled coil domain containing protein 1| Spindle apparatus coiled coil protein 1| Spindly| Spindly homolog
- AA409762| Arsenite related gene 1 protein| Arsenite-related gene 1 protein| CCDC 99| Ccdc99| Coiled coil domain containing 99| Coiled-coil domain-containing protein 99| FLJ20364| FLJ40690| hSpindly| MGC124977| Protein Spindly| rhabdomyosarcoma antigen MU RMS 40.4A| Rhabdomyosarcoma antigen MU-RMS-40.4A| Rrhabdomyosarcoma antigen protein MU RMS 40.4A| SPDL1| SPDLY_HUMAN| Spindle apparatus coiled coil domain containing protein 1| Spindle apparatus coiled coil protein 1| Spindly| Spindly homolog
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96EA4
- Q96EA4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together