Anti-CYP27A1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: CYP27A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77840-483
- 77840-482
- Antigen Symbol:
- CYP27A1
- CYP27A1
- Antigen Name:
- Sterol 26-hydroxylase, mitochondrial protein (290-407AA)
- Sterol 26-hydroxylase, mitochondrial protein (290-407AA)
- Antigen Synonyms:
- 12-alpha-triol 27-hydroxylase|5-beta-cholestane-3-alpha|7-alpha|12-alpha-triol 26-hydroxylase|5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol 27-hydroxylase|Cholestanetriol 26 monooxygenase|CP27|CP27A_HUMAN|CTX|CYP|CYP27|CYP27A1|Cytochrome P 450C27/25|Cytochrome P-450C27/25|Cytochrome P450 27|Cytochrome P450 27 mitochondrial|Cytochrome P450 family 27 subfamily A member 1|Cytochrome P450 family 27 subfamily A polypeptide 1|Cytochrome P450 subfamily XXVIIA (steroid 27-hydroxylase cerebrotendinous xanthomatosis) polypeptide 1|mitochondrial|Sterol 26 hydroxylase|Sterol 26 hydroxylase mitochondrial|Sterol 26-hydroxylase|Sterol 27 hydroxylase|Sterol 27-hydroxylase|Vitamin D(3) 25 hydroxylase|Vitamin D(3) 25-hydroxylase
- 12-alpha-triol 27-hydroxylase|5-beta-cholestane-3-alpha|7-alpha|12-alpha-triol 26-hydroxylase|5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol 27-hydroxylase|Cholestanetriol 26 monooxygenase|CP27|CP27A_HUMAN|CTX|CYP|CYP27|CYP27A1|Cytochrome P 450C27/25|Cytochrome P-450C27/25|Cytochrome P450 27|Cytochrome P450 27 mitochondrial|Cytochrome P450 family 27 subfamily A member 1|Cytochrome P450 family 27 subfamily A polypeptide 1|Cytochrome P450 subfamily XXVIIA (steroid 27-hydroxylase cerebrotendinous xanthomatosis) polypeptide 1|mitochondrial|Sterol 26 hydroxylase|Sterol 26 hydroxylase mitochondrial|Sterol 26-hydroxylase|Sterol 27 hydroxylase|Sterol 27-hydroxylase|Vitamin D(3) 25 hydroxylase|Vitamin D(3) 25-hydroxylase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02318
- Q02318
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together