Anti-FOXD4/FOXD4L4/FOXD4L5/FOXD4L3/FOXD4L6 Rabbit Polyclonal Antibody
77808-117
New Product
- Antigen Name:Synthesized peptide derived from the Internal region of FoxD4/D4L.
- Antigen Symbol:FOXD4/FOXD4L4/FOXD4L5/FOXD4L3/FOXD4L6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q12950/Q8WXT5/Q5VV16/Q6VB85/Q6VB84/Q3SYB3
- Antigen Synonyms:FOXD4| FKHL9| FOXD4A| FREAC5| Forkhead box protein D4| Forkhead-related protein FKHL9| Forkhead-related transcription factor 5| FREAC-5| Myeloid factor-alpha| FOXD4L4| FOXD4B| Forkhead box protein D4-like 4| FOXD4-like 4| Forkhead box prote
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:40000
Type:
Antigen: FOXD4/FOXD4L4/FOXD4L5/FOXD4L3/FOXD4L6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human