Order Entry
United States
ContactUsLinkComponent
Anti-E2F5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-E2F5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-E2F5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77825-449
  • 77825-450
  • Antigen Symbol:
  • E2F5
  • E2F5
  • Antigen Name:
  • Transcription factor E2F5 protein (193-298AA)
  • Transcription factor E2F5 protein (193-298AA)
  • Antigen Synonyms:
  • E2F 5|E2F transcription factor 5|E2F transcription factor 5 p130 binding|E2F-5|E2f5|E2F5_HUMAN|p130 binding|Transcription factor E2F5
  • E2F 5|E2F transcription factor 5|E2F transcription factor 5 p130 binding|E2F-5|E2f5|E2F5_HUMAN|p130 binding|Transcription factor E2F5
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q15329
  • Q15329
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together