Anti-RAB27A Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAB27A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-353
- 77796-352
- Antigen Symbol:
- RAB27A
- RAB27A
- Antigen Name:
- Ras-related protein Rab-27A protein (2-221AA)
- Ras-related protein Rab-27A protein (2-221AA)
- Antigen Synonyms:
- GS2|GTP-binding protein Ram|HsT18676|MGC117246|Mutant Ras related protein Rab-27A|Rab-27|RAB-27A|RAB27|RAB27A|RAB27A member RAS oncogene family|RAM|Ras-related protein Rab-27A|Ras-related protein Rab27A|RB27A_HUMAN
- GS2|GTP-binding protein Ram|HsT18676|MGC117246|Mutant Ras related protein Rab-27A|Rab-27|RAB-27A|RAB27|RAB27A|RAB27A member RAS oncogene family|RAM|Ras-related protein Rab-27A|Ras-related protein Rab27A|RB27A_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51159
- P51159
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together