Anti-DNAH12 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DNAH12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-151
- 77830-152
- Antigen Symbol:
- DNAH12;DHC3;DLP12;DNAH12L;DNAH7L;DNAHC3;DNHD2;HDHC3;HL19Dynein heavy chain 12;axonemal;Axonemal beta dynein heavy chain 12;Axonemal dynein heavy chain 12-like protein;Axonemal dynein heavy chain 7-like protein;Ciliary dynein heavy chain 12;Dynein heavy chain 7-like;Dynein heavy chain domain-containing protein 2
- DNAH12;DHC3;DLP12;DNAH12L;DNAH7L;DNAHC3;DNHD2;HDHC3;HL19Dynein heavy chain 12;axonemal;Axonemal beta dynein heavy chain 12;Axonemal dynein heavy chain 12-like protein;Axonemal dynein heavy chain 7-like protein;Ciliary dynein heavy chain 12;Dynein heavy chain 7-like;Dynein heavy chain domain-containing protein 2
- Antigen Name:
- Dynein heavy chain 12, axonemal protein (2423-2632AA)
- Dynein heavy chain 12, axonemal protein (2423-2632AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ZR08
- Q6ZR08
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together