Anti-DPYSL3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: DPYSL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77809-102
- 77809-103
- Antigen Symbol:
- DPYSL3
- DPYSL3
- Antigen Name:
- Fusion protein of DPYSL3
- Fusion protein of DPYSL3
- Antigen Synonyms:
- Collapsin response mediator protein 4| Collapsin response mediator protein 4 long| CRMP 4| CRMP-4| CRMP4| Dihydropyrimidinase like 3| Dihydropyrimidinase related protein 3| Dihydropyrimidinase-related protein 3| DPYL3_HUMAN| DPYSL3| DRP 3| DRP-3| DRP3| LCRMP| TUC4| ULIP 1| ULIP| ULIP-1| ULIP1| UNc 33 like phosphoprotein 1| Unc 33 like phosphoprotein| Unc-33-like phosphoprotein 1
- Collapsin response mediator protein 4| Collapsin response mediator protein 4 long| CRMP 4| CRMP-4| CRMP4| Dihydropyrimidinase like 3| Dihydropyrimidinase related protein 3| Dihydropyrimidinase-related protein 3| DPYL3_HUMAN| DPYSL3| DRP 3| DRP-3| DRP3| LCRMP| TUC4| ULIP 1| ULIP| ULIP-1| ULIP1| UNc 33 like phosphoprotein 1| Unc 33 like phosphoprotein| Unc-33-like phosphoprotein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14195
- Q14195
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together