Anti-MKKS Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, IHC:1:50 to 1:200
Type:
Antigen: MKKS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77820-470
- 77820-471
- Antigen Symbol:
- MKKS
- MKKS
- Antigen Name:
- Fusion protein of MKKS
- Fusion protein of MKKS
- Antigen Synonyms:
- Bardet Biedl syndrome 6 protein|Bardet-Biedl syndrome 6 protein|BBS6|HMCS|KMS|McKusick Kaufman syndrome|McKusick Kaufman/Bardet Biedl syndromes putative chaperonin|McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin|Mkks|MKKS_HUMAN|MKS
- Bardet Biedl syndrome 6 protein|Bardet-Biedl syndrome 6 protein|BBS6|HMCS|KMS|McKusick Kaufman syndrome|McKusick Kaufman/Bardet Biedl syndromes putative chaperonin|McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin|Mkks|MKKS_HUMAN|MKS
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NPJ1
- Q9NPJ1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together