Anti-PVRIG Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: PVRIG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77828-351
- 77828-350
- Antigen Symbol:
- PVRIG
- PVRIG
- Antigen Name:
- Transmembrane protein PVRIG protein (193-326AA)
- Transmembrane protein PVRIG protein (193-326AA)
- Antigen Synonyms:
- Poliovirus receptor-related immunoglobulin domain-containing protein|PVRIG|PVRIG_HUMAN|Transmembrane protein PVRIG
- Poliovirus receptor-related immunoglobulin domain-containing protein|PVRIG|PVRIG_HUMAN|Transmembrane protein PVRIG
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6DKI7
- Q6DKI7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together