Anti-EME1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: EME1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-935
- 77833-936
- Antigen Symbol:
- EME1
- EME1
- Antigen Name:
- Crossover junction endonuclease EME1 protein (1-583AA)
- Crossover junction endonuclease EME1 protein (1-583AA)
- Antigen Synonyms:
- 6820428D13|Crossover junction endonuclease EME1|EC 3.1.22.-|EME1|Eme1 essential meiotic endonuclease 1 homolog 1 (S. pombe)|S. pombe|homolog of|1|EME1_HUMAN|essential meiotic endonuclease 1 homolog 1 (S pombe)|Essential Meiotic Endonuclease 1 Homolog 1|essential meiotic endonuclease 1 homolog 2|Essential meiotic endonuclease 1|FLJ31364|hMMS4|homolog of yeast EME1 endonuclease|MGC106543|MMS4|MMS4 homolog|MMS4L
- 6820428D13|Crossover junction endonuclease EME1|EC 3.1.22.-|EME1|Eme1 essential meiotic endonuclease 1 homolog 1 (S. pombe)|S. pombe|homolog of|1|EME1_HUMAN|essential meiotic endonuclease 1 homolog 1 (S pombe)|Essential Meiotic Endonuclease 1 Homolog 1|essential meiotic endonuclease 1 homolog 2|Essential meiotic endonuclease 1|FLJ31364|hMMS4|homolog of yeast EME1 endonuclease|MGC106543|MMS4|MMS4 homolog|MMS4L
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96AY2
- Q96AY2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together