Anti-SLC27A4 Rabbit Polyclonal Antibody
Catalog # 77799-963
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Solute carrier family 27 member 4
- Antigen Symbol:SLC27A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q6P1M0
- Antigen Synonyms:SLC27A4|ACSVL4|FATP4|Long-chain fatty acid transport protein 4|FATP-4|Fatty acid transport protein 4|EC 6.2.1.-|Solute carrier family 27 member 4
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SLC27A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse