Anti-CD59 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: CD59
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77820-479
- 77820-478
- Antigen Symbol:
- CD59
- CD59
- Antigen Name:
- Synthetic peptide of CD59
- Synthetic peptide of CD59
- Antigen Synonyms:
- 16.3A5| 1F5 | 1F5 antigen| 20 kDa homologous restriction factor| CD 59| CD_antigen=CD59| CD59| CD59 antigen| CD59 antigen complement regulatory protein | CD59 antigen p18 20 | CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5| EJ16| EJ30| EL32 and G344)| CD59 glycoprotein| CD59 molecule| CD59 molecule complement regulatory protein| CD59_HUMAN| Cd59a| Complement regulatory protein| EL32| FLJ38134| FLJ92039| G344| HRF 20| HRF-20| HRF20| Human leukocyte antigen MIC11 | Ly 6 like protein | Lymphocytic antigen CD59/MEM43 | MAC inhibitory protein| MAC IP| MAC-inhibitory protein| MAC-IP| MACIF| MACIP| MEM43| MEM43 antigen| Membrane attack complex (MAC) inhibition factor | Membrane attack complex inhibition factor| Membrane inhibitor of reactive lysis| MGC2354| MIC11| MIN1| MIN2| MIN3| MIRL| MSK21| p18 20| Protectin| Surface antigen recognized by monoclonal 16.3A5| T cell activating protein
- 16.3A5| 1F5 | 1F5 antigen| 20 kDa homologous restriction factor| CD 59| CD_antigen=CD59| CD59| CD59 antigen| CD59 antigen complement regulatory protein | CD59 antigen p18 20 | CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5| EJ16| EJ30| EL32 and G344)| CD59 glycoprotein| CD59 molecule| CD59 molecule complement regulatory protein| CD59_HUMAN| Cd59a| Complement regulatory protein| EL32| FLJ38134| FLJ92039| G344| HRF 20| HRF-20| HRF20| Human leukocyte antigen MIC11 | Ly 6 like protein | Lymphocytic antigen CD59/MEM43 | MAC inhibitory protein| MAC IP| MAC-inhibitory protein| MAC-IP| MACIF| MACIP| MEM43| MEM43 antigen| Membrane attack complex (MAC) inhibition factor | Membrane attack complex inhibition factor| Membrane inhibitor of reactive lysis| MGC2354| MIC11| MIN1| MIN2| MIN3| MIRL| MSK21| p18 20| Protectin| Surface antigen recognized by monoclonal 16.3A5| T cell activating protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P13987
- P13987
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together