Anti-Mcpt4 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Mcpt4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77815-567
- 77815-566
- Antigen Symbol:
- Mcpt4
- Mcpt4
- Antigen Name:
- Recombinant Mouse Mast cell protease 4 protein (21-246AA)
- Recombinant Mouse Mast cell protease 4 protein (21-246AA)
- Antigen Synonyms:
- Mast cell protease 4| Mcp 4| Mcpt4| MCPT4_MOUSE| MMCP 4| MMCP 4A| MMCP 4B| MMCP-4| MMCP4| MMCP4A| MMCP4B| MSMCP| Myonase| Serosal mast cell protease
- Mast cell protease 4| Mcp 4| Mcpt4| MCPT4_MOUSE| MMCP 4| MMCP 4A| MMCP 4B| MMCP-4| MMCP4| MMCP4A| MMCP4B| MSMCP| Myonase| Serosal mast cell protease
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21812
- P21812
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together