Anti-VANGL1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: VANGL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77827-360
- 77827-361
- Antigen Symbol:
- VANGL1
- VANGL1
- Antigen Name:
- Fusion protein of VANGL1
- Fusion protein of VANGL1
- Antigen Synonyms:
- ANGL planar cell polarity protein 1|KAI1 C-terminal interacting tetraspanin|KITENIN|Loop tail protein 2 homolog|Loop-tail protein 2 homolog|LPP2|MGC5338|STB2|STBM2|Strabismus 2|Strabismus|Drosophila|homolog of|2|Van Gogh like protein 1|Van Gogh|1|Van Gogh-like protein 1|Vang like 1 (van gogh|Drosophila)|Vang like protein 1|Vang-like 1|Vang-like protein 1|VANG1_HUMAN|VANGL 1|Vangl1
- ANGL planar cell polarity protein 1|KAI1 C-terminal interacting tetraspanin|KITENIN|Loop tail protein 2 homolog|Loop-tail protein 2 homolog|LPP2|MGC5338|STB2|STBM2|Strabismus 2|Strabismus|Drosophila|homolog of|2|Van Gogh like protein 1|Van Gogh|1|Van Gogh-like protein 1|Vang like 1 (van gogh|Drosophila)|Vang like protein 1|Vang-like 1|Vang-like protein 1|VANG1_HUMAN|VANGL 1|Vangl1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TAA9
- Q8TAA9
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together