Anti-CHAC1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CHAC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-697
- 77841-698
- Antigen Symbol:
- CHAC1
- CHAC1
- Antigen Name:
- Glutathione-specific gamma-glutamylcyclotransferase 1 protein (43-264AA)
- Glutathione-specific gamma-glutamylcyclotransferase 1 protein (43-264AA)
- Antigen Synonyms:
- Blocks Notch protein|Botch|Cation transport regulator like protein 1|Cation transport regulator-like protein 1|ChaC|ChaC cation transport regulator homolog 1|ChaC cation transport regulator like 1|chac1|CHAC1_HUMAN|Gamma GCT acting on glutathione homolog 1|Gamma-GCG 1|Glutathione-specific gamma-glutamylcyclotransferase 1
- Blocks Notch protein|Botch|Cation transport regulator like protein 1|Cation transport regulator-like protein 1|ChaC|ChaC cation transport regulator homolog 1|ChaC cation transport regulator like 1|chac1|CHAC1_HUMAN|Gamma GCT acting on glutathione homolog 1|Gamma-GCG 1|Glutathione-specific gamma-glutamylcyclotransferase 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BUX1
- Q9BUX1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together