Anti-BTF3L4 Rabbit Polyclonal Antibody
77810-489
New Product
- Antigen Name:Synthesized peptide derived from the Internal region of BTF3L4.
- Antigen Symbol:BTF3L4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:Q96K17
- Antigen Synonyms:BTF3L4|Transcription factor BTF3 homolog 4|Basic transcription factor 3-like 4
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: BTF3L4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse