Anti-PARP10 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: PARP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-837
- 77832-836
- Antigen Symbol:
- PARP10
- PARP10
- Antigen Name:
- Synthetic peptide of PARP10
- Synthetic peptide of PARP10
- Antigen Synonyms:
- PARP10|Protein mono-ADP-ribosyltransferase PARP10|EC 2.4.2.-|ADP-ribosyltransferase diphtheria toxin-like 10|ARTD10|Poly [ADP-ribose] polymerase 10|PARP-10
- PARP10|Protein mono-ADP-ribosyltransferase PARP10|EC 2.4.2.-|ADP-ribosyltransferase diphtheria toxin-like 10|ARTD10|Poly [ADP-ribose] polymerase 10|PARP-10
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53GL7
- Q53GL7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together