Anti-PCSK9 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PCSK9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77810-575
- 77810-574
- Antigen Symbol:
- PCSK9
- PCSK9
- Antigen Name:
- Proprotein convertase subtilisin/kexin type 9 protein (462-595AA)
- Proprotein convertase subtilisin/kexin type 9 protein (462-595AA)
- Antigen Synonyms:
- Convertase subtilisin/kexin type 9 preproprotein|FH3|HCHOLA3|Hypercholesterolemia autosomal dominant 3|LDLCQ1|NARC 1|NARC-1|NARC1|Neural apoptosis regulated convertase 1|Neural apoptosis-regulated convertase 1|PC 9|PC9|PCSK 9|PCSK9_HUMAN|Proprotein convertase 9|Proprotein convertase PC9|Proprotein convertase subtilisin/kexin type 9|PSEC0052|Subtilisin/kexin like protease PC9|Subtilisin/kexin-like protease PC9
- Convertase subtilisin/kexin type 9 preproprotein|FH3|HCHOLA3|Hypercholesterolemia autosomal dominant 3|LDLCQ1|NARC 1|NARC-1|NARC1|Neural apoptosis regulated convertase 1|Neural apoptosis-regulated convertase 1|PC 9|PC9|PCSK 9|PCSK9_HUMAN|Proprotein convertase 9|Proprotein convertase PC9|Proprotein convertase subtilisin/kexin type 9|PSEC0052|Subtilisin/kexin like protease PC9|Subtilisin/kexin-like protease PC9
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NBP7
- Q8NBP7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together