Anti-PTPN18 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PTPN18
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-626
- 77834-627
- Antigen Symbol:
- PTPN18
- PTPN18
- Antigen Name:
- Tyrosine-protein phosphatase non-receptor type 18 protein (1-210AA)
- Tyrosine-protein phosphatase non-receptor type 18 protein (1-210AA)
- Antigen Synonyms:
- BDP 1|Brain derived phosphatase|Brain-derived phosphatase|Protein tyrosine phosphatase non receptor type 18|Protein tyrosine phosphatase non receptor type 18 brain derived|PTN18_HUMAN|PTP HSCF|PTPN 18|PTPN18|Tyrosine protein phosphatase non receptor type 18|Tyrosine-protein phosphatase non-receptor type 18
- BDP 1|Brain derived phosphatase|Brain-derived phosphatase|Protein tyrosine phosphatase non receptor type 18|Protein tyrosine phosphatase non receptor type 18 brain derived|PTN18_HUMAN|PTP HSCF|PTPN 18|PTPN18|Tyrosine protein phosphatase non receptor type 18|Tyrosine-protein phosphatase non-receptor type 18
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99952
- Q99952
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together