Anti-PIK3CG Rabbit Polyclonal Antibody
77806-178
New Product
- Antigen Name:Synthesized peptide derived from the Internal region of PI 3-Kinase p110?.
- Antigen Symbol:PIK3CG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P48736
- Antigen Synonyms:1 phosphatidylinositol 3 kinase|5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma|5-bisphosphate 3-kinase catalytic subunit gamma isoform|p110 gamma|p110gamma|p120 PI3K|p120-PI3K|Phosphatidylinositol 3 kinase catalytic 110 kD gamma|Phosphatidylinositol 3 kinase gamma, p110 gamma|Phosphatidylinositol 3 kinase, catalytic, gamma polypeptide|Phosphatidylinositol 4 5 bisphosphate 3 kinase 110 kDa catalytic subunit gamma|Phosphatidylinositol 4 5 bisphosphate 3 kinase catalytic subunit gamma|Phosphatidylinositol 4 5 bisphosphate 3 kinase catalytic subunit gamma isoform|Phosphatidylinositol-4|Phosphoinositide 3 kinase catalytic gamma polypeptide|Phosphoinositide 3 kinase gamma catalytic subunit|PI 3 Kinase catalytic subunit gamma|PI3 kinase p110 subunit gamma|PI3-kinase subunit gamma|PI3CG|PI3K|PI3K-gamma|PI3Kgamma|PIK3|Pik3cg|PK3CG_HUMAN|PtdIns-3-kinase subunit gamma|PtdIns-3-kinase subunit p110-gamma|Serine/threonine protein kinase PIK3CG
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: PIK3CG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human