Anti-SH3BGRL Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SH3BGRL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-466
- 77799-467
- Antigen Symbol:
- SH3BGRL
- SH3BGRL
- Antigen Name:
- SH3 domain-binding glutamic acid-rich-like protein (1-114AA)
- SH3 domain-binding glutamic acid-rich-like protein (1-114AA)
- Antigen Synonyms:
- MGC 117402|OTTHUMP 00000023609|SH3 binding domain glutamic acid rich protein like|SH3 domain binding glutamic acid rich protein like|SH3 domain-binding glutamic acid-rich-like protein|SH3BGR|SH3BGRL|SH3L1_HUMAN
- MGC 117402|OTTHUMP 00000023609|SH3 binding domain glutamic acid rich protein like|SH3 domain binding glutamic acid rich protein like|SH3 domain-binding glutamic acid-rich-like protein|SH3BGR|SH3BGRL|SH3L1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75368
- O75368
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together