Anti-PDCD4 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:1000 to 1:5000, IP:1:200 to 1:2000
Type:
Antigen: PDCD4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77827-507
- 77827-508
- Antigen Symbol:
- PDCD4
- PDCD4
- Antigen Name:
- Programmed cell death protein 4 protein (1-260AA)
- Programmed cell death protein 4 protein (1-260AA)
- Antigen Synonyms:
- Death up-regulated gene protein| Dug| H731| Ma3| MGC33046| MGC33047| Neoplastic transformation inhibitor| Neoplastic transformation inhibitor protein| Nuclear antigen H731| Nuclear antigen H731 like| Nuclear antigen H731 like protein| Nuclear antigen H731-like| PDCD 4| Pdcd4| PDCD4_HUMAN| Programmed cell death 4 | programmed cell death 4 (neoplastic transformation inhibitor)| Programmed cell death protein 4| Protein 197/15a| Protein MA-3| Tis| Topoisomerase-inhibitor suppressed protein
- Death up-regulated gene protein| Dug| H731| Ma3| MGC33046| MGC33047| Neoplastic transformation inhibitor| Neoplastic transformation inhibitor protein| Nuclear antigen H731| Nuclear antigen H731 like| Nuclear antigen H731 like protein| Nuclear antigen H731-like| PDCD 4| Pdcd4| PDCD4_HUMAN| Programmed cell death 4 | programmed cell death 4 (neoplastic transformation inhibitor)| Programmed cell death protein 4| Protein 197/15a| Protein MA-3| Tis| Topoisomerase-inhibitor suppressed protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53EL6
- Q53EL6
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together