Anti-HTR3D Rabbit Recombinant Polyclonal Antibody
Catalog # 77830-189
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:5-hydroxytryptamine receptor 3D protein (107-210AA)
- Antigen Symbol:HTR3D
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q70Z44
- Antigen Synonyms:HTR3D5-hydroxytryptamine receptor 3D|5-HT3-D|5-HT3D|Serotonin receptor 3D
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IF: 1:50 to 1:200
Type:
Antigen: HTR3D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human