Anti-CACNA2D1 Rabbit Polyclonal Antibody
77809-231
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:Synthesized peptide derived from the N-terminal region of Cacna2d1.
- Antigen Symbol:CACNA2D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P54289
- Antigen Synonyms:CA2D1_HUMAN| CACN A2| CACNA2| Cacna2d1| CACNL2A| Calcium channel L type alpha 2 polypeptide| Calcium channel voltage dependent alpha 2/delta subunit 1| CCHL2A| Dihydropyridine receptor alpha 2 subunit| Dihydropyridine sensitive L type calcium channel alpha 2/delta subunit| Dihydropyridine sensitive L type calcium channel subunits alpha 2/delta| L type calcium channel subunit alpha 2| MHS 3| MHS3| Voltage dependent calcium channel subunit alpha 2/delta 1| Voltage gated calcium channel subunit alpha 2/delta 1| Voltage-dependent calcium channel subunit delta-1| Voltage-gated calcium channel subunit alpha-2/delta-1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000
Type:
Antigen: CACNA2D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat