Anti-FOXD1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:500 to 1:5000, IHC: 1:10 to 1:50
Type:
Antigen: FOXD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77814-126
- 77814-125
- Antigen Symbol:
- FOXD1
- FOXD1
- Antigen Name:
- Synthetic peptide of FOXD1
- Synthetic peptide of FOXD1
- Antigen Synonyms:
- Brain Factor 2| FKH L8| FKHL 8| FKHL8| Forkhead (Drosophila) like 8| Forkhead box D1| Forkhead box protein D1| Forkhead drosophila homolog like 8| Forkhead like 8| Forkhead related activator 4| Forkhead related protein FKHL8| Forkhead related transcription factor 4| Forkhead-related protein FKHL8| Forkhead-related transcription factor 4| FOX D1| FOXD 1| foxd1| FOXD1_HUMAN| FREAC 4| FREAC-4| FREAC4
- Brain Factor 2| FKH L8| FKHL 8| FKHL8| Forkhead (Drosophila) like 8| Forkhead box D1| Forkhead box protein D1| Forkhead drosophila homolog like 8| Forkhead like 8| Forkhead related activator 4| Forkhead related protein FKHL8| Forkhead related transcription factor 4| Forkhead-related protein FKHL8| Forkhead-related transcription factor 4| FOX D1| FOXD 1| foxd1| FOXD1_HUMAN| FREAC 4| FREAC-4| FREAC4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16676
- Q16676
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together