Anti-RBL2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RBL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-560
- 77821-559
- Antigen Symbol:
- RBL2
- RBL2
- Antigen Name:
- Retinoblastoma-like protein 2 protein (417-616AA)
- Retinoblastoma-like protein 2 protein (417-616AA)
- Antigen Synonyms:
- 130 kDa retinoblastoma associated protein|130 kDa retinoblastoma-associated protein|IB4|p130|PPAR-alpha-interacting complex protein 128|PRB2|Rb2|Rbl2|RBL2_HUMAN|RBR 2|RBR-2|RBR2|Retinoblastoma like 2|Retinoblastoma-like protein 2|Retinoblastoma-related protein 2|RIC128
- 130 kDa retinoblastoma associated protein|130 kDa retinoblastoma-associated protein|IB4|p130|PPAR-alpha-interacting complex protein 128|PRB2|Rb2|Rbl2|RBL2_HUMAN|RBR 2|RBR-2|RBR2|Retinoblastoma like 2|Retinoblastoma-like protein 2|Retinoblastoma-related protein 2|RIC128
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q08999
- Q08999
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together